MilliporeSigma
  • Antiviral activity of trichothecene mycotoxins (deoxynivalenol, fusarenon-X, and nivalenol) against herpes simplex virus types 1 and 2.
Antiviral activity of trichothecene mycotoxins (deoxynivalenol, fusarenon-X, and nivalenol) against herpes simplex virus types 1 and 2.
Microbiology and immunology (1995-01-01)
N Tani, Y Dohi, Y Onji, K Yonemasu
ABSTRACT
The effect of trichothecene mycotoxins, deoxynivalenol (DON), fusarenon-X (FX) and nivalenol (NIV), on plaque formation of herpes simplex virus types 1 and 2 (HSV-1 and HSV-2) in HEp-2 cells was examined. The 50% effective concentrations (EC50) of DON, FX, and NIV for HSV-1 plaque formation were 160, 56, and 120 ng/ml, respectively. Those for HSV-2 plaque formation were 94, 26, and 50 ng/ml, respectively. These three mycotoxins showed about 2-fold higher selectivity to HSV-2 than to HSV-1. Plaque formation of HSV-1 was not inhibited with trichothecenes at concentrations completely inhibiting plaque formation when cells were treated during virus adsorption period or 15 hr before infection. These results indicate that trichothecenes affect replication of HSV-1 after virus adsorption, but not before or during virus adsorption to the host cells.
Product Description
Supelco
Fusarenon X, analytical standard
Supelco
Fusarenon X solution, 100 μg/mL in acetonitrile, analytical standard