MilliporeSigma
  • A simplified procedure for the preparation of 2,3-O-isopropylidene-sn-glycerol from L-arabinose.
A simplified procedure for the preparation of 2,3-O-isopropylidene-sn-glycerol from L-arabinose.
Journal of lipid research (1980-02-01)
P Kanda, M A Wells
ABSTRACT
A new procedure for the preparation of 2,3-O-isopropylidene-sn-glycerol is described. L-arabinose is converted to its 4,5-monoisopropylidene diethyl mercaptal derivative. This compound is then subjected to periodate oxidation and borohydride reduction. Following neutralization, the aceton-glycerol is extracted from the aqueous solution into chloroform. Evaporation of the chloroform and subsequent distillation yielded pure 2,3-O-isopropylidene-sn-glycerol ([alpha]D22 = -14.5 degrees (in substance)) in an overall yield of 15-25%.
Sigma-Aldrich
DL-1,2-Isopropylideneglycerol, ≥97.0%
Sigma-Aldrich
(S)-(+)-1,2-Isopropylideneglycerol, 98%, optical purity ee: 99% (GLC)
Sigma-Aldrich
(R)-(−)-2,2-Dimethyl-1,3-dioxolane-4-methanol, 98%