MilliporeSigma
The metabolism of 1,3-dibromopropane by the rat.
Xenobiotica; the fate of foreign compounds in biological systems (1981-08-01)
A R Jones, G Wells
ABSTRACT
1. The metabolism of 1,3-dibromopropane had been investigated in the rat. Two conjugated metabolites have been isolated from the urine and identified as S-(3-hydroxypropyl)cysteine and N-acetyl-S-(3-hydroxypropyl)cysteine. 2. An oxidation product, identified as beta-bromolactic acid, has been isolated as a urinary metabolite. 3. 1,3-dibromopropane is not excreted unchanged in expired air or in the urine. Approx. 15% of the dose (100 mg/kg) is excreted as metabolic products over 50 h and 3.5% as CO2 within 6 h, indicating that oxidation is the main route of detoxication.
Sigma-Aldrich
1,3-Dibromopropane, ReagentPlus®, 99%