MilliporeSigma
  • Metabolism of 1-0-[1'-14C]octadecyl-sn-glycerol in Leishmania donovani promastigotes. Ether lipid synthesis and degradation of the ether bond.
Metabolism of 1-0-[1'-14C]octadecyl-sn-glycerol in Leishmania donovani promastigotes. Ether lipid synthesis and degradation of the ether bond.
Molecular and biochemical parasitology (1982-02-01)
H O Herrmann, G Gercken
ABSTRACT
A long-chain 0-alkylglycerol, 1-0-[1'-14C]octadecyl-sn-glycerol, was taken up and metabolized extensively in Leishmania donovani promastigotes grown on a lipid-free, semi-defined medium as well as on a lipid-free, synthetic medium in nearly identical ways. Cleavage of the ether bond was the main event even after short incubation times (1 h) and low precursor concentration (2 microM), as judged from the appearance of radioactive acyl moieties residing mainly in phosphatidylcholine. In addition, labeled plasmanyl inositol and plasmenyl ethanolamine were formed to a considerable extent. However, when administered in higher concentrations (25 microM), 1-0-alkyl-sn-glycerols as well as their enantiomers inhibited lipid metabolism dramatically, accompanied by rounding of the cells and appearance of cellular debris in the medium. This effect was not dependent on the aliphatic side chain (12:0, 16:0, 18:0, 18:1) of the 0-alkylglycerols. It is suggested that, by incorporation into cellular membranes, 0-alklglycerols affect factors responsible for the interaction of membranes and the cytoskeleton as well as enzymes of lipid metabolism.
Sigma-Aldrich
Batyl alcohol, 99%