MilliporeSigma
  • Problems of the use of 2,2-dimethoxypropane as dehydrating agent in preparing single cells for transmission electron microscopy.
Problems of the use of 2,2-dimethoxypropane as dehydrating agent in preparing single cells for transmission electron microscopy.
Histochemistry (1982-01-01)
R Dierichs, C Dosche
ABSTRACT
Dehydration with 2,2-dimethoxypropane acidified with 1 n HCl in an 1:100 volume ratio has been found to be unsuitable for the preservation of cell membranes in single cell preparations. Reducing the HCl concentration to 0.1 or 0.01 n results in an improvement of membrane structures. This effect is rather independent of different fixation modes, buffer concentrations, or embedding media. It is suggested that membrane destruction is caused by a proton concentration exceeding a suitable level.
Product Description
Supelco
2,2-Dimethoxypropane, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
2,2-Dimethoxypropane, reagent grade, 98%
Pricing and availability is not currently available.