MilliporeSigma
  • Comparative studies of the hepatic effects of di- and mono-n-octyl phthalates, di-(2-ethylhexyl) phthalate and clofibrate in the rat.
Comparative studies of the hepatic effects of di- and mono-n-octyl phthalates, di-(2-ethylhexyl) phthalate and clofibrate in the rat.
Acta pharmacologica et toxicologica (1984-03-01)
B G Lake, W R Rijcken, T J Gray, J R Foster, S D Gangolli
ABSTRACT
The oral administration of di-n-octyl phthalate (DNOP), mono-n-octyl phthalate (MNOP), di-(2-ethylhexyl) phthalate (DEHP) and clofibrate to young male Sprague-Dawley rats for 14 days resulted in liver enlargement. Morphological examination of liver sections from DEHP and clofibrate treated rats, but not from either DNOP or MNOP treated animals, revealed increased numbers of peroxisomes (microbodies). Both DEHP and clofibrate treatment markedly stimulated the activities of certain peroxisomal marker enzymes whereas DNOP and MNOP produced only marginal effects. Similarly both DEHP and clofibrate, but not DNOP or MNOP, increased microsomal cytochrome P-450 content and markedly stimulated microsomal lauric acid hydroxylation activity. The results thus demonstrate that whilst the branched chain phthalate ester DEHP induced peroxisomal proliferation, the straight chain analogue DNOP and its metabolite MNOP were essentially inactive. In addition, DEHP treatment appeared to induce similar form(s) of cytochrome P-450 in rat liver to those previously described after clofibrate administration.
Product Description
Supelco
Di-n-octyl phthalate, analytical standard
Sigma-Aldrich
Di-n-octyl phthalate, ≥98.0% (GC)
Supelco
Di-n-octyl phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland