MilliporeSigma
  • Influence of phenobarbital and 3-methylcholanthrene on the metabolism of aminopyrine in isolated hepatocyte system.
Influence of phenobarbital and 3-methylcholanthrene on the metabolism of aminopyrine in isolated hepatocyte system.
Journal of pharmacobio-dynamics (1983-11-01)
K Matsuyama, A Noda, S Goto, T Tanaka, S Iguchi
ABSTRACT
The effect of phenobarbital (PB) and 3-methylcholanthrene (3-MC) on the metabolic behavior of aminopyrine (AM) was studied using an isolated hepatocyte system prepared from male Wistar rats. The formation of 4-formylaminoantipyrine (FAA) was increased after pretreatment with PB, but not 3-MC. However, the total amounts of AM and its main metabolites recovered from hepatocyte system after the incubation for 30 min were considerably reduced both by PB and 3-MC-pretreatment. Furthermore, when using 4-monomethylaminoantipyrine (MAA), the first metabolite of AM, as a substrate, 3-MC-pretreatment resulted in a more significant decrease in the total amounts of MAA and its further metabolites recovered than did PB. Present observation suggests the participation of cytochrome P-448 as well as cytochrome P-450 in the metabolism of AM.
Product Description
Supelco
4-Formylaminoantipyrine, VETRANAL®, analytical standard
Metamizole impurity A, European Pharmacopoeia (EP) Reference Standard