MilliporeSigma
  • Oxidation and dehalogenation of 4-chlorophenylacetate by a two-component enzyme system from Pseudomonas sp. strain CBS3.
Oxidation and dehalogenation of 4-chlorophenylacetate by a two-component enzyme system from Pseudomonas sp. strain CBS3.
Journal of bacteriology (1984-11-01)
A Markus, U Klages, S Krauss, F Lingens
ABSTRACT
In cell-free extracts from Pseudomonas sp. strain CBS3 the conversion of 4-chlorophenylacetate to 3,4-dihydroxyphenylacetate was demonstrated. By Sephacryl S-200 chromatography two protein fractions, A and B, were obtained which both were essential for enzyme activity. Fe2+ and NADH were cofactors of the reaction. NADPH also activated the enzyme, but less effectively than NADH. FAD had no influence on enzyme activity. 4-Hydroxyphenylacetate, 4-chloro-3-hydroxyphenylacetate, and 3-chloro-4-hydroxyphenylacetate were poor substrates for the enzyme, suggesting that these substances are not intermediates of the reaction. We therefore suggest that the reaction proceeds via a dioxygenated intermediate.
Sigma-Aldrich
4-Chlorophenylacetic acid, ReagentPlus®, 99%