MilliporeSigma
  • Pulmonary and percutaneous absorption of 2-propoxyethyl acetate and 2-ethoxyethyl acetate in beagle dogs.
Pulmonary and percutaneous absorption of 2-propoxyethyl acetate and 2-ethoxyethyl acetate in beagle dogs.
Environmental health perspectives (1984-08-01)
D Guest, M L Hamilton, P J Deisinger, G D DiVincenzo
ABSTRACT
A comparison was made of the absorption and elimination rates of 2-propoxyethyl acetate (PEA) and 2-ethoxyethyl acetate (EEA) following inhalation, dermal application or IV administration. Male beagle dogs were exposed to 50 ppm PEA or EEA for 5 hr, and breath samples were collected during the exposure and a 3-hr recovery period. Both compounds were rapidly absorbed through the lungs. After 10 min of exposure, the concentrations of the parent compounds in the expired breath were 5 to 10 ppm (80-90% absorption) and reached plateau values at about 3 hr of 13 ppm for PEA (74% absorption) and 16 ppm for EEA (68% absorption). Post-exposure breath samples declined exponentially to 0.5 ppm and 2 ppm after 3 hr for PEA and EEA, respectively. Expired concentrations of PEA were slightly, but significantly (p less than 0.025), lower than those of EEA at corresponding times during the exposure. After IV dosing with 1 mg/kg [ethyl-1,2-14C]PEA, the urine contained 61% and 88% of the dose in 4 and 24 hr, respectively. [14C]EEA was eliminated more slowly, with 20% and 61% of the dose appearing in the urine in 4 and 24 hr, respectively. Blood elimination half-lives were 1.6 hr for [14C]PEA and 7.9 hr for [14C]EEA. Only trace amounts of 14CO2 (less than 1%) or volatile materials (less than 0.1%) were detected in the expired air with either compound. For studies of percutaneous absorption, [14C]PEA or [14C]EEA was added to undiluted compound and applied in a glass cell to a shaved area on a dog's thorax for 30 or 60 min.(ABSTRACT TRUNCATED AT 250 WORDS)
Sigma-Aldrich
2-Ethoxyethyl acetate, 98%