MilliporeSigma
[Allergy to dyes in stockings].
Deutsche medizinische Wochenschrift (1946) (1984-09-28)
B M Hausen, K H Schulz
ABSTRACT
Skin allergies caused by the wearing of stockings and hose have received little attention. Findings in patients of an allergy department, enquiries at stocking counters of stores and recent publications indicate, however, that probably many more persons have an allergy to stocking dyes than is generally thought. Skin tests with isolated stocking dyes indicate that azo dye dispersion yellow 3, dispersion orange 3 and dispersion red 1 are the most important contact allergens. They were demonstrated in 18-21 of the 23 hose examined. In textile materials, azo dye dispersion blue 124 is predominant among allergens. Cross-reactions may occur to other dispersion azo dyes, used in cosmetics, textiles, toiletries and hygenic articles, permitted food additives and hair dyes. It is suggested that in persons who have dye allergy or intolerance, decolouration followed by colouring with natural colours be undertaken.
Sigma-Aldrich
Disperse Yellow 3, Dye content 30 %
Supelco
Disperse Yellow 3, analytical standard