MilliporeSigma
  • Lesions induced in rodent pancreas by azaserine and other pancreatic carcinogens.
Lesions induced in rodent pancreas by azaserine and other pancreatic carcinogens.
Environmental health perspectives (1984-06-01)
D S Longnecker
ABSTRACT
Focal proliferative changes in the acinar cells of the pancreas of rats have been induced by several systemically administered carcinogens including azaserine, N-nitrosobis(2-oxopropyl)amine, N-nitroso-(2-hydroxypropyl) (2-oxopropyl)amine, and N delta-(N-methyl-N-nitrosocarbamoyl)-L-ornithine (MNCO). Foci, nodules, and adenomas induced by these carcinogens are usually made up of atypical-appearing acinar cells that maintain a high degree of differentiation, but a minority of these lesions exhibit anaplastic cellular changes that suggest the development of malignant potential. Such anaplasia may occupy the whole of smaller lesions or may occur as a secondary focal change within larger nodules or adenomas. Many foci and nodules per pancreas have been induced by single or multiple exposures to these known genotoxic carcinogens, but relatively few of them develop into carcinomas. Azaserine and MNCO have induced acinar cell carcinomas in rats. Those induced by azaserine have exhibited a broad spectrum of histologic variants, including ductlike cystic, and undifferentiated patterns. Higher doses of MNCO have induced a second pattern of change in the pancreatic lobules of rats, which includes cystic and tubular ductlike structures that have been called cystic and tubular ductal complexes. MNCO has also induced focal acinar cell lesions, cystic and tubular ductal complexes, and adenocarcinomas in the pancreas of Syrain golden hamsters. In this species, ductal complexes are much more numerous than are proliferative lesions of acinar cells, and the histologic appearance of the carcinomas is ductlike. Hyperplasia and atypical changes were also seen in the epithelium of the intralobular ducts of hamsters.(ABSTRACT TRUNCATED AT 250 WORDS)
Product Description
Supelco
N-Nitrosodi-n-propylamine, analytical standard
Supelco
N-Nitrosodi-n-propylamine solution, certified reference material, 5000 μg/mL in methanol