MilliporeSigma
  • Alternate substrates of dopamine beta-hydroxylase. III. Stoichiometry of the inactivation reaction with benzyl cyanides and spectroscopic investigations.
Alternate substrates of dopamine beta-hydroxylase. III. Stoichiometry of the inactivation reaction with benzyl cyanides and spectroscopic investigations.
The Journal of biological chemistry (1984-02-10)
G Colombo, B Rajashekhar, D E Ash, J J Villafranca
ABSTRACT
Dopamine beta-hydroxylase was incubated with p-hydroxybenzyl cyanide, ascorbate, and O2 and the products of the hydroxylation reaction were monitored by high performance liquid chromatography. At early times, p-hydroxymandelonitrile was the sole product but this compound slowly decomposed to p-hydroxybenzaldehyde and cyanide. Dopamine beta-hydroxylase was also inhibited under these reaction conditions but the amount of cyanide produced was insufficient to account for the extent of enzyme inhibition. Incubation of dopamine beta-hydroxylase with [ring-3H]p-hydroxybenzyl cyanide, ascorbate, and O2 resulted in incorporation of detectable radiolabel into enzyme only when 100% O2 (0.4 X Km) was present. The amount of incorporated radiolabel was substoichiometric with respect to enzyme subunits (approximately 30%) and also did not correlate with the total amount of inhibited dopamine beta-hydroxylase. These data suggest two different modes of inhibition: covalent-adduct formation at early times and inhibition due to cyanide generated at the active site. Separate K14CN binding studies with dopamine beta-hydroxylase corroborated the latter suggestion since 2 mol of cyanide were bound per tetramer. Tyramine stabilized [14C]cyanide binding to dopamine beta-hydroxylase, consistent with the lack of enzyme reactivation observed under certain conditions (Colombo, G., Giedroc, D. P., Rajashekhar, B., and Villafranca, J. J. 1984) J. Biol. Chem. 259, 1601-1606). EPR data for dopamine beta-hydroxylase-bound Cu2+ directly demonstrated that a quaternary complex with tyramine and cyanide was formed, since the spectrum of dopamine beta-hydroxylase-Cu2+-tyramine-cyanide is distinct from that of dopamine beta-hydroxylase-Cu2+-cyanide and dopamine beta-hydroxylase-Cu2+-tyramine. A comprehensive mechanism for dopamine beta-hydroxylase inhibition by benzyl cyanides is presented.
Sigma-Aldrich
4-Hydroxyphenylacetonitrile, 98%