MilliporeSigma
Allergic contact dermatitis from an organic di-isocyanate.
Contact dermatitis (1983-07-01)
I R White, J R Stewart, R J Rycroft
ABSTRACT
Organic di-isocyanates are used widely in industry. The concomitant respiratory complications are well-known. Cutaneous problems precipitated by skin contact with these chemicals are less well recognised. Allergic contact dermatitis caused by exposure to di-cyclohexylmethane 4,4'-di-isocyanate has been demonstrated in a factory where it was used to form a surface-coating polyurethane. Attention to hygiene methods may have reduced the incidence of cases.
Product Description
Supelco
HMDI, analytical standard
Sigma-Aldrich
4,4′-Methylenebis(cyclohexyl isocyanate), mixture of isomers, 90%