MilliporeSigma
  • Para-chlorophenylethylamine in brains of rats treated with a monoamine oxidase inhibitor and p-chlorophenylalanine.
Para-chlorophenylethylamine in brains of rats treated with a monoamine oxidase inhibitor and p-chlorophenylalanine.
Progress in neuro-psychopharmacology & biological psychiatry (1982-01-01)
G B Baker, I L Martin, R T Coutts, A Benderly
ABSTRACT
1. The accumulation of p-chlorophenylethylamine (pCPE) in rat brain after administration of pargyline plus p-chlorophenylalanine (pCPA) is demonstrated. 2. Measurements of pCPE in brain were performed at 0.25 h, 1 h and 4 h after administration of pCPA to pargyline-pretreated rats, and amounts of pCPE were shown to increase with time.
Sigma-Aldrich
2-(4-Chlorophenyl)ethylamine, 98%