MilliporeSigma
  • The deamination of n-pentylamine by monoamine oxidase and a semicarbazide-sensitive amine oxidase of rat heart.
The deamination of n-pentylamine by monoamine oxidase and a semicarbazide-sensitive amine oxidase of rat heart.
The Journal of pharmacy and pharmacology (1983-07-01)
C Guffroy, C J Fowler, M Strolin Benedetti
ABSTRACT
n-Pentylamine is deaminated by homogenates of rat heart. Clorgyline inhibition curves at 10 and 100 microM n-pentylamine indicated that this substrate was deaminated by MAO-A, -B and a clorgyline-resistant amine oxidase sensitive to inhibition by semicarbazide. These results have been compared with two other commonly used monoamine substrates, beta-phenethylamine and benzylamine.
Sigma-Aldrich
Amylamine, ≥99%, FG
Sigma-Aldrich
Amylamine, 99%