MilliporeSigma
Treatment of recurrent earlobe keloids.
Cutis (1986-01-01)
G E Rauscher, W L Kolmer
ABSTRACT
The use of complete surgical excision and postoperative pressure with steroid-impregnated tape has been evaluated in fifty-seven patients with recurrent earlobe keloids. All patients underwent conventional treatment previously, which was not curative. In a four-year follow-up of the method described here, four recurrences occurred. Patients' failure to use the recommended pressure earring and alteration in endocrine balance were noted in three of the four patients with recurrent keloids.