MilliporeSigma
  • Phenyl-n-butylborinic acid is a potent transition state analog inhibitor of lipolytic enzymes.
Phenyl-n-butylborinic acid is a potent transition state analog inhibitor of lipolytic enzymes.
Biochemical and biophysical research communications (1986-01-14)
L D Sutton, J S Stout, L Hosie, P S Spencer, D M Quinn
ABSTRACT
The cholesterol esterase and lipoprotein lipase catalyzed hydrolyses of the water-soluble substrate p-nitrophenyl butyrate are competitively inhibited by butaneboronic acid and phenylboronic acid. Phenyl-n-butylborinic acid has been synthesized and characterized as an ultrapotent transition state analog inhibitor: Ki = 2.9 +/- 0.6 nM and 1.7 +/- 0.3 microM for the cholesterol esterase and lipoprotein lipase reactions, respectively. These results are interpreted in terms of transition state structure and stabilization.
Product Description
Supelco
Butylboronic acid, for GC derivatization, LiChropur, ≥96.0% (T)
Sigma-Aldrich
Butylboronic acid, 97%