MilliporeSigma
  • Metabolism of 2- and 3-tert-butyl-4-hydroxyanisole (2- and 3-BHA) in the rat (I): Excretion of BHA in urine, feces and expired air and distribution of BHA in the main organs.
Metabolism of 2- and 3-tert-butyl-4-hydroxyanisole (2- and 3-BHA) in the rat (I): Excretion of BHA in urine, feces and expired air and distribution of BHA in the main organs.
Toxicology (1987-02-01)
M Hirose, A Hagiwara, K Inoue, T Sakata, N Ito, H Kaneko, A Yoshitake, J Miyamoto
ABSTRACT
The mechanism of the carcinogenic or toxic action of BHA on rat forestomach was examined by studies on the excretion and tissue distribution of radioactivity in F344 male rats given tert-butyl- or methoxy-labelled 3-BHA orally. Within 2 days after a single oral dose of labelled BHA at 1 g/kg body wt, 87-96% of the 14C was excreted, mainly in the urine with smaller amounts in the feces and expired air. More 14C was found in the tissues of rats given the methoxy-labelled compounds. The distributions of 14C in the forestomach and the glandular stomach were similar. At 168 h after treatment, more 14C was found in the forestomach of rats given 2-BHA than in that of rats given 3-BHA. These results indicate that excretion of BHA is rapid, that 4-O-methyl demethylation may take place readily and that demethylated methyl group may become distributed non-specifically in tissues. The carcinogenic or toxic action of BHA on the forestomach does not seem to be due accumulation of BHA in the forestomach.
Sigma-Aldrich
3-tert-Butyl-4-hydroxyanisole, ≥98% (sum of isomers, GC), ≤10% 2-BHA basis (GC)
USP
2-tert-Butyl-4-hydroxyanisole, United States Pharmacopeia (USP) Reference Standard