MilliporeSigma
  • Tumorigenic activity of non-alternant polynuclear aromatic hydrocarbons in newborn mice.
Tumorigenic activity of non-alternant polynuclear aromatic hydrocarbons in newborn mice.
Cancer letters (1987-01-01)
E J Lavoie, J Braley, J E Rice, A Rivenson
ABSTRACT
The tumorigenic activity of benzo[b]fluoranthene, benzo[j]fluoranthene, benzo[k]fluoranthene, and indeno-[1,2,3-cd]pyrene was evaluated in newborn CD-1 mice. The total doses of these non-alternant polycyclic aromatic hydrocarbons employed in this study ranged from 0.5 to 2.1 mumol per mouse. The results of this assay indicate that both benzo[b]fluoranthene and benzo[j]fluoranthene exhibit significant tumorigenic activity. In contrast to these results, neither benzo[k]fluoranthene nor indeno[1,2,3-cd]pyrene were tumorigenic under these assay conditions.
Product Description
Supelco
Indeno[1,2,3-cd]pyrene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Indeno[1,2,3-cd]pyrene, analytical standard
Supelco
Benzo[j]fluoranthene solution, certified reference material, 2000 μg/mL in dichloromethane