MilliporeSigma
  • Use of high-performance liquid chromatography in the measurement of in vitro acetylation in man.
Use of high-performance liquid chromatography in the measurement of in vitro acetylation in man.
Journal of chromatography (1986-02-26)
J D Baty, R M Lindsay, S Sharp
ABSTRACT
Liquid chromatographic methods were developed for the study of the in vitro acetylation of the sulphonamide drug sulphamethazine and a series of aniline derivatives. The sensitivity of the methods have allowed data on the activity of the N-acetyltransferase enzyme(s) in man to be obtained. The use of bonded-phase columns with a series of organic mobile phases has been compared with reversed-phase separation of the acetanilide derivatives, produced in the acetylation reaction.
Sigma-Aldrich
4-Ethoxyaniline, 98%