MilliporeSigma
  • Indirect influence of s-triazines on rat gonadotropic mechanism at early postnatal period.
Indirect influence of s-triazines on rat gonadotropic mechanism at early postnatal period.
Journal of steroid biochemistry (1987-01-01)
J Kniewald, M Peruzović, T Gojmerac, K Milković, Z Kniewald
ABSTRACT
Daily s.c. injections of atrazine and deethylatrazine to rat mothers during pregnancy only or during pregnancy and lactation influenced the pituitary-gonadal axis of male and female offsprings. In female and male offspring, slow maturation of gonadotropic system is evident and as a consequence modified male and female pituitary 5 alpha-R activity is present. The number of specific steroid-hormone receptor sites at the offsprings' gonads is unchanged if the mothers were treated only during pregnancy, but 5 alpha-DHT prostate receptors are strongly inhibited in offspring of mothers treated with atrazine and deethylatrazine during pregnancy and lactation.
Product Description
Supelco
Atrazine-desethyl, PESTANAL®, analytical standard