MilliporeSigma
  • Mutagenicity of chloroolefins in the Salmonella/mammalian microsome test. III. Metabolic activation of the allylic chloropropenes allyl chloride, 1,3-dichloropropene, 2,3-dichloro-1-propene, 1,2,3-trichloropropene, 1,1,2,3-tetrachloro-2-propene and hexachloropropene by S9 mix via two different metabolic pathways.
Mutagenicity of chloroolefins in the Salmonella/mammalian microsome test. III. Metabolic activation of the allylic chloropropenes allyl chloride, 1,3-dichloropropene, 2,3-dichloro-1-propene, 1,2,3-trichloropropene, 1,1,2,3-tetrachloro-2-propene and hexachloropropene by S9 mix via two different metabolic pathways.
Mutation research (1986-04-01)
T Neudecker, D Henschler
ABSTRACT
In the presence of S9 mix all allylic chloropropenes tested exert considerable indirect mutagenic activity which is most pronounced for 1,2,3-trichloropropene. Lower as well as higher chlorinated derivatives are clearly less mutagenic. Longer than standard incubation time (120 min instead of 20 min) at 37 degrees C always leads to an increase in mutagenic activity. An increase in concentration of rat-liver homogenate fraction (S9) in the metabolising system (S9 mix) enhances mutagenicity only for 1,3-dichloropropene, 2,3-dichloro-1-propene and for the cis isomer of 1,1,2,3-tetrachloro-2-propene. According to the effects of the enzyme inhibitors SKF525 1,1,1-trichloropropene-2,3-oxide and cyanamide the allylic chloropropenes fall into 3 groups distinguished by their mode of metabolic activation by S9 mix: (a) allyl chloride and 1,3-dichloropropene are hydrolysed to the corresponding allylic alcohols which can be oxidised to the respective acroleins (hydrolytic-oxidative pathway); (b) 2,3-dichloro-1-propene, 1,1,2,3-tetrachloro-2-propene and hexachloropropene are epoxidised in the C=C double bond, giving rise to reactive epoxides (epoxidative pathway); (c) only 1,2,3-trichloropropene is obviously activated by both these alternative metabolic pathways. Structural parameters like chloro-substitution of the central C atom of the C=C-C sequence and substituent-induced polarisation of the C=C double bond as well as cis/trans isomerism might be responsible for different substrate properties for the enzymes involved in allylic chloropropene metabolism, thus determining different degrees of activation by either one or both pathways.
Sigma-Aldrich
Hexachloropropene, ≥90%
Sigma-Aldrich
2,3-Dichloro-1-propene, 98%