MilliporeSigma
  • Bacterial metabolism of side chain fluorinated aromatics: cometabolism of 4-trifluoromethyl(TFM)-benzoate by 4-isopropylbenzoate grown Pseudomonas putida JT strains.
Bacterial metabolism of side chain fluorinated aromatics: cometabolism of 4-trifluoromethyl(TFM)-benzoate by 4-isopropylbenzoate grown Pseudomonas putida JT strains.
Archives of microbiology (1988-01-01)
K H Engesser, M A Rubio, D W Ribbons
ABSTRACT
Enzymes of the p-cymene pathway in Pseudomonas putida strains cometabolized the intermediate analogue 4-trifluoromethyl(TFM)benzoate. Three products, 4-TFM-2,3-dihydro-2,3-dihydroxybenzoate, 4-TFM-2,3-dihydroxybenzoate and 2-hydroxy-6-oxo-7,7,7-trifluorohepta-2,4-dienoate (7-TFHOD) were identified chemically and by spectroscopic properties. Certain TFM-substituted analogue metabolites of the p-cymene pathway were transformed at drastically reduced rates. Hammett type analysis of ring cleavage reactions of 4-substituted 2,3-dihydroxybenzoates revealed the negative inductive and especially mesomeric effect of substituents to be rate determining. Whereas decarboxylation of 3-carboxy-7-TFHOD was not affected by fluorine substitution the subsequent hydrolysis of 7-TFHOD proceeded very slowly. The negative inductive effect of the TFM-group probably inhibited heterolysis of the carbon bond between C5 and C6 of 7-TFHOD.
Sigma-Aldrich
4-(Trifluoromethyl)benzoic acid, 98%
Sigma-Aldrich
4-Isopropylbenzoic acid, ≥96%