MilliporeSigma
Metabolism of 2,6-dimethylnaphthalene by flavobacteria.
Applied and environmental microbiology (1988-02-01)
E A Barnsley
ABSTRACT
Flavobacteria that were able to grow on 2,6-dimethylnaphthalene (2,6-DMN) were isolated from soil. Most were able to oxidize a broad range of aromatic hydrocarbons after growth on 2,6-DMN at rates comparable to that of the oxidation of 2,6-DMN itself. One small group was neither able to grow on naphthalene nor able to oxidize this compound after growth on 2,6-DMN, but metabolized 2,6-DMN by a pathway which converged with that previously described for naphthalene metabolism in pseudomonads. These organisms could also grow on salicylate or methylsalicylate, and in so doing, early enzymes for 2,6-DMN metabolism were induced.
Sigma-Aldrich
2,6-Dimethylnaphthalene, 99%