MilliporeSigma
  • New type of pullulanase from Bacillus stearothermophilus and molecular cloning and expression of the gene in Bacillus subtilis.
New type of pullulanase from Bacillus stearothermophilus and molecular cloning and expression of the gene in Bacillus subtilis.
Journal of bacteriology (1988-04-01)
T Kuriki, S Okada, T Imanaka
ABSTRACT
A new type of pullulanase which mainly produced panose from pullulan was found in Bacillus stearothermophilus and purified. The enzyme can hydrolyze pullulan efficiently and only hydrolyzes a small amount of starch. When pullulan was used as a substrate, the main product was panose and small amounts of glucose and maltose were simultaneously produced. By using pTB522 as a vector plasmid, the enzyme gene was cloned and expressed in Bacillus subtilis. Since the enzyme from the recombinant plasmid carrier could convert pullulan into not only panose but also glucose and maltose, we concluded that these reactions were due to the single enzyme. The new pullulanase, with a molecular weight of 62,000, was fairly thermostable. The optimum temperature was 60 to 65 degrees C, and about 90% of the enzyme activity was retained even after treatment at 60 degrees C for 60 min. The optimum pH for the enzyme was 6.0.