MilliporeSigma
  • Specific tritium labeling of glucosyl- and galactosylceramides at the 6-position of the carbohydrate moiety using CrO3-graphite.
Specific tritium labeling of glucosyl- and galactosylceramides at the 6-position of the carbohydrate moiety using CrO3-graphite.
Analytical biochemistry (1986-01-01)
S Usuki, Y Nagai
ABSTRACT
A new procedure for introducing tritium into the carbohydrate portions of glucosyl- and galactosylceramides was developed using a new catalyst, CrO3-graphite, which specifically oxidizes the primary alcohol group to the aldehyde. About 10% of the glycolipid was converted to the aldehyde and the aldehyde produced was then reduced back to the original form with KB3H4. After methanolysis, more than 96.7% of the radioactivities of [3H]glucosyl- and [3H]galactosylceramides were found to be located in the carbohydrate portions, and the specific activities of the [3H]galactosyl- and [3H]glucosylceramides were 2.08 to 4.30 X 10(4) cpm/nmol, which could be increased greatly by purifying the aldehydes and reducing them with KB3H4. In addition, beta-galactosidase activity was successfully determined with [3H]galactosylceramide as the enzyme substrate; the Km was 18.73 mM and the Vmax was 11.63 nmol/mg/h, indicating that no significant structural modification occurs during the oxidation.
Sigma-Aldrich
Potassium borohydride, 99.9% trace metals basis
Sigma-Aldrich
Potassium borohydride, purum, ≥97.0% (RT)
Sigma-Aldrich
Potassium borohydride, ≥97%