MilliporeSigma
  • Ion-pair reverse-phase high-performance liquid chromatography. Application to the study of chicken liver NAD+ kinase.
Ion-pair reverse-phase high-performance liquid chromatography. Application to the study of chicken liver NAD+ kinase.
Analytical biochemistry (1985-09-01)
M K Gabriel, E T McGuinness
ABSTRACT
An ion-pair, reverse-phase, high-performance liquid chromatography method of assay was developed and used in a series of rate studies carried out with the enzyme chicken liver NAD+ kinase (ATP:NAD+ 2'-phosphotransferase, EC 2.7.1.23). Complete separation of all products and reactants was achieved within 15 min. ATP, NAD+, ADP, and NADP+ were monitored at 260 nm as they eluted from a Zorbax (Dupont) ODS (4.6 X 250-mm) column using an acetonitrile and 0.01 mM NH4(H2PO4)/0.005 M tetrabutylammonium phosphate (pH 7.0) gradient. The enzyme shows a marked preference for ATP (and dATP) and Mg2+ (or Mn2+) relative to other trinucleotides and divalent metal ions. It exhibits residual adenylate kinase and ATPase activity, but no NADH kinase activity. When polyphosphate replaced ATP, NADP+ production dropped to 2.5%. The addition of Ca2+ and/or bovine brain calmodulin did not significantly enhance the rate of NADP+ production.
Sigma-Aldrich
Polyphosphoric acid, ~105% H3PO4 basis
Sigma-Aldrich
Polyphosphoric acid, reagent grade, 115% H3PO4 basis