MilliporeSigma
Inhibition of hydroxymethylglutaryl-coenzyme A synthase by L-659,699.
Proceedings of the National Academy of Sciences of the United States of America (1987-11-01)
M D Greenspan, J B Yudkovitz, C Y Lo, J S Chen, A W Alberts, V M Hunt, M N Chang, S S Yang, K L Thompson, Y C Chiang
ABSTRACT
A beta-lactone isolated from Fusarium sp. has been shown to be a potent specific inhibitor of the enzyme 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) synthase [(S)-3-hydroxy-3-methylglutaryl-CoA acetoacetyl-CoA-lyase (CoA-acetylating), EC, 4.1.3.5] from rat liver. The structure of this beta-lactone, termed L-659,699, is (E,E)-11-[3-(hydroxy-methyl)-4-oxo-2-oxytanyl]-3,5,7-trimethyl-2,4 - undecadienenoic acid. A partially purified preparation of cytoplasmic HMG-CoA synthase from rat liver was inhibited by L-659,699 with an IC50 of 0.12 microM. The enzyme HMG-CoA reductase, beta-ketoacyl-CoA thiolase, acetoacetyl-CoA synthetase, and fatty acid synthase were not inhibited to any extent by this compound. In cultured Hep G2 cells, the compound inhibited the incorporation of [14C]acetate into sterols with an IC50 of 6 microM, while incorporation of [3H]mevalonate into sterols in these cells was not affected. The activity of HMG-CoA reductase in the cultured Hep G2 cells was induced in a dose-dependent manner by incubation with L-659,699. A 37-fold increase in reductase was observed after a 24-hr incubation with 62 microM L-659,699. The effect of a number of analogs of L-659,699 on HMG-CoA synthase is also discussed.
Sigma-Aldrich
Hymeglusin, ≥98% (HPLC), from Fusarium sp.