MilliporeSigma
  • Covalent binding to apoprotein is a major fate of heme in a variety of reactions in which cytochrome P-450 is destroyed.
Covalent binding to apoprotein is a major fate of heme in a variety of reactions in which cytochrome P-450 is destroyed.
Biochemical and biophysical research communications (1986-07-16)
F P Guengerich
ABSTRACT
The heme in rat liver microsomal cytochrome P-450 was labeled with 14C or 3H and the microsomes were fractionated after in vitro incubations with a variety of agents known to destroy cytochrome P-450 heme. A major fraction of the heme label was irreversibly bound to apoprotein in all cases, including incubations with fluroxene, 1-octene, vinyl bromide, trichloroethylene, vinyl chloride, parathion, cumene hydroperoxide, NaN3, or iron-ADP complex. Label was also extensively bound to apoprotein when purified and reconstituted cytochrome P-450 was incubated with NADPH and vinyl chloride. This process appears to be widespread and involved to a significant extent in the cytochrome P-450 heme destruction observed with many compounds.
Sigma-Aldrich
1-Octene, 98%
Supelco
1-Octene, analytical standard