MilliporeSigma
  • Determination of absolute chemiluminescence quantum yields for reactions of bis-(pentachlorophenyl) oxalate, hydrogen peroxide and fluorescent compounds.
Determination of absolute chemiluminescence quantum yields for reactions of bis-(pentachlorophenyl) oxalate, hydrogen peroxide and fluorescent compounds.
Journal of bioluminescence and chemiluminescence (1989-07-01)
C L Catherall, T F Palmer, R B Cundall
ABSTRACT
Absolute chemiluminescence quantum yields (phi CL) for reactions of bis-(pentachlorophenyl) oxalate (PCPO), hydrogen peroxide (H2O2) and 9:10 diphenyl anthracene (DPA) have been determined. A fully corrected chemiluminescence monitoring spectrometer was calibrated for spectral sensitivity using the chemiluminescence of the bis-(pentachlorophenyl) oxalate system as a liquid light source, the total photon output of which had previously been determined by chemical actinometry. At high (PCPO)/(H2O2) ratios phi CL was found to be independent of PCPO and H2O2 concentrations.
Sigma-Aldrich
9,10-Diphenylanthracene, 97%
Supelco
9,10-Diphenylanthracene, analytical standard