MilliporeSigma
  • Derivatization of N-methyl and cyclic amino acids with dimethylformamide dimethyl acetal.
Derivatization of N-methyl and cyclic amino acids with dimethylformamide dimethyl acetal.
Journal of chromatography (1989-05-19)
M F Grubb, P S Callery
ABSTRACT
Six amino acids containing either an N-methyl or a cyclic secondary amine were converted to volatile derivatives by reaction with dimethylformamide dimethyl acetal. The amine functionalities were formylated by way of an amide acetal intermediate while the carboxylic acid groups were esterified directly. The resulting N-formyl esters were stable to solvent extraction and exhibited gas chromatography-mass spectrometry properties suitable for assay development.
Sigma-Aldrich
N,N-Dimethylformamide dimethyl acetal, technical grade, 94%
Supelco
N,N-Dimethylformamide dimethyl acetal, for GC derivatization, LiChropur