MilliporeSigma
  • Factors affecting the response of lung clearance systems to acid aerosols: role of exposure concentration, exposure time, and relative acidity.
Factors affecting the response of lung clearance systems to acid aerosols: role of exposure concentration, exposure time, and relative acidity.
Environmental health perspectives (1989-02-01)
R B Schlesinger
ABSTRACT
The ability of the lungs to clear deposited material is essential for maintenance of lung homeostasis. Acid aerosols have been shown to alter the efficiency of this process. This paper assesses the role of acid aerosol exposure concentration (C), exposure time (T), and relative acidity in producing changes in clearance from both the tracheobronchial tree and respiratory region of the lungs of rabbits. The response was found to be due to total exposure, i.e., some combination form of C x T, and was also related to relative acidity.
Sigma-Aldrich
Ammonium hydrogensulfate, 99.99% trace metals basis