MilliporeSigma
Detection of a Myc-associated protein by chemical cross-linking.
Molecular and cellular biology (1989-02-01)
D A Gillespie, R N Eisenman
ABSTRACT
A single nuclear protein (Myc-associated protein) can be specifically cross-linked to avian Myc proteins by treatment of nuclei or cells with the reversible cross-linker dimethyl 3,3'-dithiobis-propionimidate. Myc-associated protein has a molecular weight of approximately 500,000, is not detectably phosphorylated and, in contrast to Myc, has a long apparent half-life of greater than 3 h.
Sigma-Aldrich
Dimethyl 3,3′-dithiopropionimidate dihydrochloride, powder