MilliporeSigma
Antimutagenicity of some citrus fruits in Salmonella typhimurium.
Mutation research (1989-03-01)
S Bala, I S Grover
ABSTRACT
The antimutagenic effect of 10 citrus fruit juices was observed against the mutagenicity of N-nitro-o-phenylenediamine (NPD) in TA97a and sodium azide in TA100 tester strains of Salmonella typhimurium using the Ames test. It was noticed that the juices of all these fruits reduced significantly the NPD and sodium azide induced revertant colonies. The inhibitory activity was enhanced if the mutagen and juice were co-incubated for about 30 min at 37 degrees C prior to performing the mutagenicity assay. Dilution with distilled water led to the reduction in the inhibitory activity. The antimutagenic activity of synthetic ascorbic acid or citric acid or combined ascorbic acid and citric acid was also seen. But the results with fruit juices tempted us to believe that in addition to ascorbic acid and citric acid, the presence of other factor(s) possessing antimutagenic properties cannot be ruled out.
Sigma-Aldrich
2-Nitro-1,4-phenylenediamine, 95%