MilliporeSigma
  • Analysis of a glucose-containing tetrasaccharide by high-performance liquid affinity chromatography.
Analysis of a glucose-containing tetrasaccharide by high-performance liquid affinity chromatography.
Analytical biochemistry (1989-10-01)
W T Wang, J Kumlien, S Ohlson, A Lundblad, D Zopf
ABSTRACT
In the present work we have explored conditions for using a pulsed amperometric detector for on-line analysis of oligosaccharides eluted from a high-performance liquid affinity chromatography column. A monoclonal antibody that specifically binds a glucose-containing oligosaccharide is coupled to a SelectiSphere-10-activated tresyl column. The system is eluted isocratically and easily detects 10 ng of the oligosaccharide with a linear response up to 250 ng. Analysis of both serum and urine samples from normal individuals and patients with acute pancreatitis gives a single retarded peak with a retention time identical to that of authentic (Glc)4. Retarded material pooled from several analyses of urine was positively identified as (Glc)4 by GC-MS analysis. As this method requires little cleanup and no chemical derivitization of the sample and is performed rapidly (less than 20 min) at sensitivities of at least 10 micrograms/liter in biological fluids, it represents a substantial improvement over previous GC-MS, radioimmunoassay, and enzyme-linked immunoadsorbent assay methods used to determine (Glc)4.
Sigma-Aldrich
Glucose tetrasaccharide, ≥99.0% (TLC)