MilliporeSigma
  • Chloride current activated by cyclic AMP and parathyroid hormone in rat osteoblasts.
Chloride current activated by cyclic AMP and parathyroid hormone in rat osteoblasts.
Pflugers Archiv : European journal of physiology (1989-10-01)
D Chesnoy-Marchais, J Fritsch
ABSTRACT
In primary cultures of rat osteoblasts, studied with the whole-cell configuration of the patch-clamp technique, 8-bromo-cyclic AMP (8BrcAMP) forskolin (FS) and 1-34 parathyroid hormone (PTH) were shown to activate a Cl conductance. This conductance shows a pronounced outward rectification, even with symmetrical Cl concentrations. It is blocked partially and reversibly by 4,4'-diisothiocyanatostilbene 2,2'-disulfonic acid (DIDS) or diphenylcarboxylate (DPC). The blockade induced by DIDS is time- and voltage-dependent. The Cl responses to FS and PTH develop slowly, after a delay of several seconds and are very slowly reversible. These responses were observed only in a fraction of the cells tested and their detection was favoured by cell dialysis. This Cl current should be taken into account for studying possible modulations of the voltage-gated Ca currents of osteoblasts. It is suggested that its physiological role may be related to the well-known morphological changes induced by PTH in osteoblasts. The cyclic AMP-sensitivity, the outward rectification and the sensitivity to dialysis of this Cl current are reminiscent of the properties of the cystic fibrosis-sensitive Cl channels of epithelial cells.
Sigma-Aldrich
Phenyl benzoate, 99%