MilliporeSigma
  • Effect of sodium hypophosphite feeding on adenosine triphosphatase activity in ageing Zaprionus paravittiger (Diptera).
Effect of sodium hypophosphite feeding on adenosine triphosphatase activity in ageing Zaprionus paravittiger (Diptera).
Gerontology (1989-01-01)
R Wadhwa, S P Sharma
ABSTRACT
Addition of antioxidant, sodium hypophosphite (SHP; 1 mM), to the diet of Zaprionus paravittiger (Diptera) results in depressed ATPase activity in both the sexes in comparison to the normal group. However, the rate of age-related decline in ATPase activity is lowered considerably in SHP fed group. Such changes in ATPase activity might be one of the important factors responsible for increased life span of flies with SHP feeding.
Sigma-Aldrich
Sodium hypophosphite, puriss., meets analytical specification of BPC63, anhydrous, 98-101%
Sigma-Aldrich
Sodium hypophosphite monohydrate, ≥99%