MilliporeSigma
  • [Identification of catecholamines in the rat brain and mollusk tissues using semithin slices and retrograde staining].
[Identification of catecholamines in the rat brain and mollusk tissues using semithin slices and retrograde staining].
Fiziologicheskii zhurnal (1989-11-01)
I M Prudnikov, V A Maĭskiĭ, E N Chepera
ABSTRACT
The suggested technique allows revealing the transport-specific dye (primulin) and catecholamine fluorescence simultaneously in the same cell of brain. Intense fluorescence is observed when brain tissue is quickly dehydrated and embedded in the epoxy resin. The same method is suggested for the identification of catecholamines in the embryonal and juvenile tissues of gastropod Lymnaea stagnalis (Mollusca Pulmonata) without using of primulin dye.
Product Description
Supelco
Malvidin-3-galactoside chloride, analytical standard