MilliporeSigma
  • [The symptomatic therapy of hemorrhoids and anal eczema--a report of experiences from proctology practice].
[The symptomatic therapy of hemorrhoids and anal eczema--a report of experiences from proctology practice].
Schweizerische Rundschau fur Medizin Praxis = Revue suisse de medecine Praxis (1990-08-02)
A Neiger, E Herms
ABSTRACT
Besides the various operative procedures, which nowadays have come into use in hemorrhoidal disorders, drug therapy as well continues to keep an important place. Its main conditions of application are inflammatory processes before, between and after hemorrhoidal sclerosis or proctological operations. In a clinical study suppositories with the corticoid fluocortolone-21-pivalate and the local anesthetic lidocaine hydrochloride as well as a cream with the same substances and in addition, chlorquinaldol were tested in 92 patients with hemorrhoidal diseases and their concomitant conditions. In 92% of the cases efficacy of the preparations proved to be good or very good, and in all cases they were well tolerated.
Sigma-Aldrich
5,7-Dichloro-8-hydroxy-2-methylquinoline, 98%