MilliporeSigma
  • Analysis of thickness of a hydrophobic fluoropolymer film based on electrowetting.
Analysis of thickness of a hydrophobic fluoropolymer film based on electrowetting.
Journal of biomedical nanotechnology (2013-08-06)
Hyojin Ko, Minh Dinh Phan, Dibyendu Khatua, Chan-Hee Jung, Jae-Hak Choi, Oh-Sun Kwon, Kwanwoo Shin
ABSTRACT
The electrowetting of water drops on a dielectric fluoropolymer film was studied experimentally. The dependence of the contact angles of the water drops on the applied voltage has been well explained in the low-voltage limit by using the classical Young-Lippmann theory. With this theory, the thicknesses of films coated on glass substrates by using a spin-coater were obtained indirectly by fitting the contact angle data and were confirmed by using X-ray reflectometry. The two sets of results showed a good agreement. In addition, we confirmed that the contact angle saturation at high voltage were consistent with Peykov's model.
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, >40 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), beads
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), 1 μm particle size
Sigma-Aldrich
Polytetrafluoroethylene preparation, 60 wt % dispersion in H2O
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 35 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), ≤12 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 200 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, ≥350 μm particle size