MilliporeSigma
  • Preparation, photoluminescence and photocatalytic properties of ZnO:Eu3+ nanocrystals.
Preparation, photoluminescence and photocatalytic properties of ZnO:Eu3+ nanocrystals.
Journal of nanoscience and nanotechnology (2013-08-02)
Fuhai Li, Hai Liu, Lixin Yu
ABSTRACT
The ZnO short nanorods and flowerlike nanostructures doped with Eu3+ ions are prepared by a hydrothermal method through adjusting the hydrothermal temperature. The XRD, FESEM, FT-IR, luminescent and photocatalytic properties are characterized. The results indicate that the hydrothermal temperature affects the morphology of ZnO:Eu3+ nanocrystals, short nanorods at 120 degrees C, flowerlike nanocrystals at 150 degrees C and nanoparticles at 180 degrees C. The structures of ZnO:Eu3+ nanocrystals prepared at different hydrothermal temperatures are assigned to hexagonal phase. The photoluminescent properties including excitation and emission spectra are studied. The charge transfer band and f-f transition absorption are observed in excitation spectra. And, the 5D(0)-7F(J) (J = 0-4) transition emissions appear in emission spectra. In addition, the photocatalytic properties are also studied. The degradation rate of ZnO:Eu3+ nanocrystals is a little lower than pure ZnO nanocrystals.
Product Description
Supelco
Zinc oxide, analytical standard
Sigma-Aldrich
Zinc oxide, 99.99% trace metals basis
Sigma-Aldrich
Zinc oxide, 99.999% trace metals basis
Sigma-Aldrich
Zinc oxide, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5% (calc. for dried substance)
Sigma-Aldrich
Zinc oxide, puriss. p.a., ACS reagent, ≥99.0% (KT)
Sigma-Aldrich
Zinc oxide, ReagentPlus®, powder, <5 μm particle size, 99.9%
Sigma-Aldrich
Zinc oxide, nanopowder, <100 nm particle size
Sigma-Aldrich
Zinc oxide, dispersion, nanoparticles, <100 nm particle size (TEM), ≤40 nm avg. part. size (APS), 20 wt. % in H2O
Sigma-Aldrich
Zinc oxide, nanopowder, <50 nm particle size (BET), >97%