MilliporeSigma
Paravertebral high cervical chordoma. A case report.
The neuroradiology journal (2013-07-19)
A Elefante, F Caranci, M L Del Basso De Caro, C Peca, E Guadagno, R Severino, G Mariniello, F Maiuri
ABSTRACT
Spinal chordomas are more often located on the midline and are associated with marked destruction of the vertebral bodies. We report a rare case of large cervical (C2-C3) right lateral paravertebral chordoma extending into the spinal canal through a very enlarged intervertebral foramen. The tumor was initially diagnosed as a mucous adenocarcinoma on a percutaneous needle biopsy. However, the neuroradiological features, including the well-defined tumor margins, the regular and sclerosing lytic bone changes with regular enlargement of the intervertebral C2-C3 foramen, were in favor of a more slowly growing lesion, such as schwannoma or neurofibroma. At surgery a well-demarcated capsulated tumor involving the nerve root was partially resected. Histology was in favor of a low-grade chordoma (Ki-67/MIB-1<1%). Postoperative proton beam therapy was also performed. The differential neuroradiological diagnosis is discussed.
Sigma-Aldrich
Keratin from human epidermis, aqueous solution