MilliporeSigma
[TiO2-DNA nanocomposites capable of penetrating into cells].
Bioorganicheskaia khimiia (2013-07-13)
A S Levina, Z R Ismagilova, M N Repkova, N V Shikina, S I Baĭborodin, N V Shatskaia, S N Zagrebel'nyĭ, V F Zarytova
ABSTRACT
Methods of noncovalent immobilization of DNA fragments onto titanium dioxide nanoparticles (TiO2) were developed, which led to TiO2-DNA nanocomposites capable of penetrating through cell membranes. TiO2 nanoparticles of different forms (amorphous, anatase, brookit) with enhanced agglomeration stability were synthesized. The particles were characterized by X-ray diffraction, small angle X-ray scattering, infrared spectroscopy and atomic force microscopy. Three approaches to the preparation of nanocomposites are described: (1) sorption of polylysine-containing oligonucleotides onto TiO2-nanoparticles, (2) the electrostatic binding of oligonucleotides to TiO2 nanoparticles bearing immobilized polylysine, and (3) sorption of oligonucleotides on TiO2 nanoparticles in the presence of cetavlon. All three methods provide an efficient and stable immobilization of DNA fragments onto nanoparticles, which leads to nanocomposites with a density for an oligonucleotide up to 40 nmol/mg. It is shown that DNA fragments in nanocomposites retain their ability to form complementary complexes and can be delivered into cells without transfection agents and other methods of exposure.
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from herring testes, Type XIV
Sigma-Aldrich
Deoxyribonucleic acid from human placenta, buffered aqueous solution, sexed, female
Sigma-Aldrich
Plasmid DNA from E. coli RRI, pUC19, buffered aqueous solution
Sigma-Aldrich
Plasmid DNA from E. coli RRI, pUC18, buffered aqueous solution
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from salmon testes, For hybridization
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from salmon testes, For hybridization