MilliporeSigma
  • Experimental study on a room temperature urea-SCR of NO over activated carbon fibre-supported CeO2-CuO.
Experimental study on a room temperature urea-SCR of NO over activated carbon fibre-supported CeO2-CuO.
Environmental technology (2013-07-11)
Xiao Jiang, Pei Lu, Caiting Li, Zheng Zeng, Guangming Zeng, Luping Hu, Lei Mai, Zhi Li
ABSTRACT
In order to establish a desirable method for NO reduction, selective catalytic reduction (SCR) of NO by urea-CeO2/ACF and urea-CeO2-CuO/ACF was carried out at room temperature. The experimental results showed that 10% urea-9% CeO2/ACF could yield the highest NO conversion of 85% among the series of urea-CeO2/ACF prepared. When urea-CeO2-CuO/ACF was compared with urea-CeO2/ACF, it achieved higher NO conversion to a certain degree with the addition of CuO, which was attributed to the synergistic effect between cerium and copper. The effect of the mass ratio of CeO2 and CuO was also observed. The desirable mass ratio of CeO2 and CuO was 1:1, which yielded about 90% NO conversion when ACF was loaded with 10% urea. Furthermore, the influence of O2 concentration and NO concentration was also observed. In this study, NO conversion increased with increasing O2 concentration. In addition, some samples were further characterized by BET, X-ray diffraction, X-ray photoelectron spectroscopy and Fourier transform infrared methods.
Product Description
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Copper(II) oxide, nanopowder, <50 nm particle size (TEM)
Continuous cast copper (O), BCR®, certified reference material, rod
Sigma-Aldrich
Copper, foil, ≥99.8% (complexometric)
Sigma-Aldrich
Copper, beads, 2-8 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 1.0 mm, ≥99.9%
Sigma-Aldrich
Copper, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 2.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.64 mm, 99.995% trace metals basis
Sigma-Aldrich
Copper(II) oxide, powder, 99.995% trace metals basis
Sigma-Aldrich
Copper, beads, 2-8 mm, 99.9995% trace metals basis
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Copper, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper(II) oxide, powder, 99.99% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, powder, <75 μm, 99%
Sigma-Aldrich
Copper(II) oxide, 99.999% trace metals basis
Sigma-Aldrich
Copper, turnings, purum p.a., ≥99.0%
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Copper(II) oxide on alumina, 14-30 mesh, extent of labeling: 13 wt. % loading
Sigma-Aldrich
Copper(II) oxide, powder, <10 μm, 98%
Sigma-Aldrich
Copper(II) oxide, needles, mixture of CuO and Cu2O, ACS reagent
Sigma-Aldrich
Copper(II) oxide, ACS reagent, ≥99.0%
Sigma-Aldrich
Copper, nanopowder, 60-80 nm particle size (SAXS), ≥99.5% trace metals basis
Sigma-Aldrich
Copper, nanopowder, 40-60 nm particle size (SAXS), ≥99.5% trace metals basis
Sigma-Aldrich
Cerium(IV) oxide, nanopowder, <50 nm particle size (BET), 99.95% trace rare earth metals basis
Sigma-Aldrich
Cerium(IV) oxide, powder, <5 μm, 99.9% trace metals basis
Sigma-Aldrich
Cerium, ingot, under oil, 99.9% trace rare earth metals basis