MilliporeSigma
[Toxicity of acrylamide and its metabolite - glicydamide].
Medycyna pracy (2013-07-09)
Daria Pingot, Kacper Pyrzanowski, Jaromir Michałowicz, Bozena Bukowska
ABSTRACT
Acrylamide is a synthetic chemical compound commonly used in many branches of industry. It is mainly used in the synthesis of polyacrylamides, which are widely employed in plastics, paints, varnishes, adhesives and mortars production. Acrylamide is also applied in the cellulose-paper and cosmetic industries to produce toiletries and cosmetics. The interest in acrylamide increased in 2002, when Swedish scientists showed that a considerable amount of this substance is formed during frying and baking of various foods. Studies concerning toxicity of acrylamide and its metabolite - glicydamide showed their neurotoxic, genotoxic and carcinogenic effects. Neverthless, in humans only neurotoxic effect of acrylamide has been clearly evidenced. Genotoxic nature of acetylamide manifests itself mainly in its metabolic conversion to the epoxide derivative glicydamide. Carcinogenic effects of acrylamide have been shown in animal studies. Epidemiological studies have not provided explicit evidence that acrylamide supplied with the diet can initiate the formation of tumors in humans. Acrylamide exposure is assessed by measuring specific compounds (adducts) formed during the reaction of acrylamide with hemoglobin and DNA.
Sigma-Aldrich
Acrylamide, for Northern and Southern blotting, powder blend
Sigma-Aldrich
Acrylamide, purum, ≥98.0% (GC)
Supelco
Acrylamide, analytical standard
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Supelco
Acrylamide, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Acrylamide, Molecular Biology, ≥99% (HPLC)
Supelco
Acrylamide solution, 40% in H2O
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered