MilliporeSigma
Lead-elevated activity of xanthine oxidase in lead-exposed workers.
Medycyna pracy (2013-07-09)
Sławomir Kasperczyk, Michał Dobrakowski, Alina Ostałowska, Aleksandra Kasperczyk, Slawomir Wilczyiński, Magdalena Wyparło-Wszelaki, Jacek Kiełtucki, Ewa Birkner
ABSTRACT
The aim of the present study was to explore the connection between lead toxicity and the activity of xanthine oxidase (XO). In addition, we indicated the uric acid (UA) and creatinine levels and concentration of erythrocyte malondialdehyde (MDA) to estimate oxidative stress intensity. The examined group consisted of 125 healthy male employees of zinc and lead works. The examined group was divided into tertiles according to blood lead levels. In the collected blood samples, concentrations of lead-exposure indices, UA, creatinine, and MDA as well as activity of XO were measured concomitantly. The control group consisted of 32 healthy male administrative workers who were exposed to lead only environmentally. XO activity and MDA level were significantly elevated in all tertiles compared to the control group. Creatinine level was significantly elevated in the medium and high tertiles. However, the level of UA was significantly elevated in the high tertile, while in the low and medium tertile only a tendency toward higher values was observed. Occupational exposure to lead induces activity of XO. 'lhis induction may contribute to the observed simultaneously increased oxidative stress, measured as MDA level, and the increased level of UA.
Sigma-Aldrich
Lead, shot, 1-3 mm, 99.995% trace metals basis
Sigma-Aldrich
Lead, shot, <2 mm, 99.9% trace metals basis
Sigma-Aldrich
Lead, powder, −100 mesh, 99.95% trace metals basis
Sigma-Aldrich
Lead, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Uric acid, ≥99%, crystalline
Sigma-Aldrich
Uric acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
Creatinine, anhydrous, ≥98%