MilliporeSigma
  • [The directed modification of Escherichia coli MG1655 to obtain histidine-producing mutants].
[The directed modification of Escherichia coli MG1655 to obtain histidine-producing mutants].
Prikladnaia biokhimiia i mikrobiologiia (2013-06-26)
V G Doroshenko, A O Lobanov, E A Fedorina
ABSTRACT
Strain MG 1655+hisGr hisL'-Delta, purR, which produces histidine with a weight yield of approximately 12% from glucose, was constructed through directed chromosomal modifications of the laboratory Escherichia coli strain MG 1655+, which has a known genome sequence. A feedback-resistant ATP-phosphoribosyl transferase encoded by the mutant hisGr (E271 K) was the main determinant of histidine production. A further increase in histidine production was achieved by the expression enhance of a mutant his operon containing hisGr through the deleting attenuator region (hisL'-Delta). An increase in the expression of the wildtype his operon did not result in histidine accumulation. Deletion of the transcriptional regulator gene purR increased the biomass produced and maintained the level of histidine production per cell under the fermentation conditions used.
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
L-Histidine, suitable for cell culture, meets EP, USP testing specifications, from non-animal source
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
L-Histidine, ReagentPlus®, ≥99% (TLC)
SAFC
L-Histidine
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Supelco
D-(+)-Glucose, analytical standard
Supelco
L-Histidine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Histidine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
L-Histidine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
D-(+)-Glucose solution, 45% in H2O, sterile-filtered, BioXtra, suitable for cell culture
Supelco
D-(+)-Glucose solution, 1 mg/mL in 0.1% benzoic acid, standard for enzymatic assay kits GAGO20, GAHK20, STA20, analytical standard
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
D-(+)-Glucose solution, 100 g/L in H2O, sterile-filtered, BioXtra, suitable for cell culture
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-(−)-Glucose, ≥99%