MilliporeSigma
  • Recycling silicon wire-saw slurries: separation of silicon and silicon carbide in a ramp settling tank under an applied electrical field.
Recycling silicon wire-saw slurries: separation of silicon and silicon carbide in a ramp settling tank under an applied electrical field.
Journal of the Air & Waste Management Association (1995) (2013-06-22)
Tzu-Hsuan Tsai, Yu-Pei Shih, Yung-Fu Wu
ABSTRACT
The growing demand for silicon solar cells in the global market has greatly increased the amount of silicon sawing waste produced each year. Recycling kerf Si and SiC from sawing waste is an economical method to reduce this waste. This study reports the separation of Si and SiC using a ramp settling tank. As they settle in an electrical field, small Si particles with higher negative charges have a longer horizontal displacement than SiC particles in a solution of pH 7, resulting in the separation of Si and SiC. The agreement between experimental results and predicted results shows that the particles traveled a short distance to reach the collection port in the ramp tank. Consequently, the time required for tiny particles to hit the tank bottom decreased, and the interference caused by the dispersion between particles and the fluid motion during settling decreased. In the ramp tank, the highest purities of the collected SiC and Si powders were 95.2 and 7.01 wt%, respectively. Using a ramp tank, the recycling fraction of Si-rich powders (SiC < 15 wt%) reached 22.67% (based on the whole waste). This fraction is greater than that achieved using rectangular tanks. Recycling Si and SiC abrasives from the silicon sawing waste is regarded as an economical solution to reduce the sawing waste. However, the separation of Si and SiC is difficult. This study reports the separation of Si and SiC using a ramp settling tank under an applied electrical field. As they settle in an electrical field, small Si particles with higher negative charges have a longer horizontal displacement than SiC particles in a solution of pH 7, resulting in the separation of Si and SiC. Compared with the rectangular tanks, the recycling fraction of Si-rich powders using a ramp tank is greater, and the proposed ramp settling tank is more suitable for industrial applications.
Sigma-Aldrich
Silicon carbide, nanopowder, <100 nm particle size
Sigma-Aldrich
Silicon carbide, -200 mesh particle size
Sigma-Aldrich
Silicon carbide, −400 mesh particle size, ≥97.5%
Sigma-Aldrich
Silicon, wafer, <111>, P-type, contains boron as dopant, diam. × thickness 2 in. × 0.3 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <111>, N-type, contains no dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, P-type, contains boron as dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, N-type, contains no dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <111>, N-type, contains no dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <111>, N-type, contains phosphorus as dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, P-type, contains boron as dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <111>, P-type, contains boron as dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, N-type, contains no dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, N-type, contains phosphorus as dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silicon, nanopowder, <100 nm particle size (TEM), ≥98% trace metals basis
Sigma-Aldrich
Silicon, pieces, 99.95% trace metals basis
Sigma-Aldrich
Silicon, powder, −325 mesh, 99% trace metals basis
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, N-type, contains phosphorus as dopant, diam. × thickness 2 in. × 0.5 mm