MilliporeSigma
  • [Experimental models of lysosomal phase reactivity in blood leukocytes exposed to low doses of potassium cyanide].
[Experimental models of lysosomal phase reactivity in blood leukocytes exposed to low doses of potassium cyanide].
Meditsina truda i promyshlennaia ekologiia (2013-06-22)
M V Dolgushin, G D Khomuev
ABSTRACT
Cytochemical analysis of acid phosphatase was used to evaluate lysosomal membranes stability under oral intake of potassium cyanide by rats over one month in daily doses of 1.30 mg/kg (1/10 LD50) and 0.65 mg/kg (1/20 LD50). The authors demonstrated phase-related dose-dependent changes in the lysosomal state, and the main response feature was associated with functional activation that usually followed the membrane alteration.
Sigma-Aldrich
Potassium cyanide-15N, 98 atom % 15N
Sigma-Aldrich
Potassium cyanide, BioUltra, ≥98.0% (AT)
Sigma-Aldrich
Potassium cyanide, technical, ≥96%
Sigma-Aldrich
Potassium cyanide, ACS reagent, ≥96.0%