MilliporeSigma
  • A prospective clinical evaluation of the longevity of resorbable sutures in oral surgical procedures.
A prospective clinical evaluation of the longevity of resorbable sutures in oral surgical procedures.
Nigerian journal of clinical practice (2013-06-19)
B Fomete, B D Saheeb, A C Obiadazie
ABSTRACT
The objectives of this prospective randomized study were to clinically evaluate the longevity of resorbable sutures (chromic catgut and vicryl) and determine the effect of chlorhexidine mouth wash on their absorption time in oral surgical procedures. Both sutures were of size 3/0 with round body needle and were placed using the standard technique. One hundred patients selected for the study were divided into two groups of 50 patients each (vicryl and chromic catgut groups). Each group was subsequently subdivided into chlorhexidine (17 cases); warm saline mouth wash (17 cases) and warm water mouth wash (16 cases (control). The sutures were placed during various minor surgical procedures e.g. third molar surgery (65 cases), incisional biopsy (7 cases), excisional biopsy (8 cases), sutured lacerations (10 cases) and malar elevations through intraoral upper buccal sulci approach (8 cases) and 2 cases of cystic enucleations. The mean longevity of chromic catgut for chlorhexidine was 11.4 days with a range of 5-16 days. The patients, who used warm saline mouth wash, had a mean longevity of 11.7 days with a range of 7-24 days. The mean longevity of vicryl in patients that used chlorhexidine was 22.7 days, with a range of 14-36 days and that of warm saline mouth wash was 24.5 days with a range of 14-47 days. The resorbable sutures investigated have a mean longevity, which was slightly shorter than the figure stated by the manufacturer. Chlorhexidine was found to have no appreciable effect on absorption time of the sutures.
Sigma-Aldrich
Resomer® RG 504 H, Poly(D,L-lactide-co-glycolide), acid terminated, lactide:glycolide 50:50, Mw 38,000-54,000
Sigma-Aldrich
Resomer® RG 505, Poly(D,L-lactide-co-glycolide), ester terminated, Mw 54,000-69,000
Sigma-Aldrich
Resomer® RG 502 H, Poly(D,L-lactide-co-glycolide), acid terminated, viscosity 0.16-0.24 dL/g 
Sigma-Aldrich
Resomer® RG 752 H, Poly(D,L-lactide-co-glycolide), acid terminated, lactide:glycolide 75:25, Mw 4,000-15,000
Sigma-Aldrich
Resomer® RG 653 H, Poly(D,L-lactide-co-glycolide), acid terminated, Mw 24,000-38,000
Sigma-Aldrich
Resomer® RG 502, Poly(D,L-Lactide-co-Glycolide), lactide:glycolide 50:50, ester terminated, Mw 7,000-17,000
Sigma-Aldrich
Resomer® RG 503 H, Poly(D,L-lactide-co-glycolide), acid terminated, lactide:glycolide 50:50, Mw 24,000-38,000
Sigma-Aldrich
Resomer® RG 504, Poly(D,L-lactide-co-glycolide), lactide:glycolide 50:50, ester terminated, Mw 38,000-54,000
Sigma-Aldrich
Resomer® RG 858 S, Poly(D,L-lactide-co-glycolide), ester terminated, lactide:glycolide 85:15, Mw 190,000-240,000
Sigma-Aldrich
Resomer® RG 503, Poly(D,L-lactide-co-glycolide), lactide:glycolide 50:50, ester terminated, Mw 24,000-38,000
Sigma-Aldrich
Resomer® RG 756 S, Poly(D,L-lactide-co-glycolide), ester terminated, lactide:glycolide 75:25, Mw 76,000-115,000
Sigma-Aldrich
Poly(D,L-lactide-co-glycolide), ester terminated, Mw 50,000-75,000
Sigma-Aldrich
Poly(D,L-lactide-co-glycolide), lactide:glycolide 65:35, Mw 40,000-75,000
Sigma-Aldrich
Poly(D,L-lactide-co-glycolide), lactide:glycolide (75:25), mol wt 66,000-107,000
Sigma-Aldrich
Chlorhexidine dihydrochloride, ≥98%
Sigma-Aldrich
Poly(D,L-lactide-co-glycolide), lactide:glycolide (50:50), mol wt 30,000-60,000
Chlorhexidine dihydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Chlorhexidine, ≥99.5%
Chlorhexidine, European Pharmacopoeia (EP) Reference Standard